Merck
Determination of acidity constants and prediction of electrophoretic separation of amyloid beta peptides.
Journal of chromatography. A (2017-06-18)
Roger Peró-Gascón, Fernando Benavente, José Barbosa, Victoria Sanz-Nebot
ABSTRACT
In this paper we describe a strategy to estimate by CE the acidity constants (pK
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Diethylmalonic acid, 98%