Merck
  • Psychological, situational and application-related determinants of the intention to self-test: a factorial survey among students.
Psychological, situational and application-related determinants of the intention to self-test: a factorial survey among students.
BMC health services research (2017-07-12)
Pinar Kuecuekbalaban, Tim Rostalski, Silke Schmidt, Holger Muehlan
ABSTRACT
The Internet enables an unprecedented opportunity to access a broad range of self-tests (e.g. testing for HIV, cancer, hepatitis B/C), which can be conducted by lay consumers without the help of a health professional. However, there is only little knowledge about the determinants of the use of self-tests. Thus, the aims of this study were (1) to experimentally investigate the impact of situational and application-related characteristics on the intention to use a self-test (ST), compared to being tested by a health professional at home (HPH) or at a doctor's office (HPD), (2) to examine the applicability of social-cognitive health behaviour theories on self-testing, and (3) to explore the advantages of integrating technological affinity into social-cognitive health behaviour models to predict self-testing. In a factorial survey, 1248 vignettes were rated by 208 students. The core concepts of social-cognitive health behaviour theories, technological affinity, and different situational and application-related characteristics were investigated. Intention to ST was only predicted by the medical expertise of the tested person, while HPH and HPD were also associated with the application purpose of the test and the presence of an emotionally supporting person. Perceived severity and outcome-expectancy significantly predicted intention to self-test. Technological enthusiastic people had a higher intention to use a self-test. Intention to ST, HPH and HPD were predicted by different situational and application-related characteristics. Social-cognitive health behaviour theories can be applied to predict self-testing and do not need to be extended by technological affinity.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
1,1,1-Trifluoro-5,5-dimethyl-2,4-hexanedione, 98%