Merck
  • Identification, characteristics and human exposure assessments of triclosan, bisphenol-A, and four commonly used organic UV filters in indoor dust collected from Shanghai, China.
Identification, characteristics and human exposure assessments of triclosan, bisphenol-A, and four commonly used organic UV filters in indoor dust collected from Shanghai, China.
Chemosphere (2017-06-18)
Junjie Ao, Tao Yuan, Yuning Ma, Li Gao, Ni Ni, Dan Li
ABSTRACT
Indoor dust is a very significant medium to understand human exposure of emerging contaminants. A novel and robust analytical method to measure the amounts of six personal care products (PCPs) (triclosan, bisphenol-A and four commonly used organic ultraviolet (UV) filters) simultaneously in indoor dust is developed in this paper. Target analytes were extracted using accelerated solvent extraction. After sample cleanup by solid-phase extraction (SPE), the extracts were derivatized and analyzed using gas chromatography-tandem mass spectrometry. The method detection limits achieves 0.16-0.62 pg g
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Chlorotrimethylsilane, purified by redistillation, ≥99%
Sigma-Aldrich
Trifluoroacetamide, 97%
Sigma-Aldrich
Benzophenone-d10, 99 atom % D