Merck
  • Gemcitabine and Chk1 Inhibitor AZD7762 Synergistically Suppress the Growth of Lkb1-Deficient Lung Adenocarcinoma.
Gemcitabine and Chk1 Inhibitor AZD7762 Synergistically Suppress the Growth of Lkb1-Deficient Lung Adenocarcinoma.
Cancer research (2017-07-30)
Yan Liu, Yuyang Li, Xiaoen Wang, Feiyang Liu, Peng Gao, Max M Quinn, Fei Li, Ashley A Merlino, Cyril Benes, Qingsong Liu, Nathanael S Gray, Kwok-Kin Wong
ABSTRACT
Cells lacking the tumor suppressor gene
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
MISSION® esiRNA, targeting human CHEK1