Merck
In vivo loss-of-function screens identify KPNB1 as a new druggable oncogene in epithelial ovarian cancer.
Proceedings of the National Academy of Sciences of the United States of America (2017-08-16)
Michiko Kodama, Takahiro Kodama, Justin Y Newberg, Hiroyuki Katayama, Makoto Kobayashi, Samir M Hanash, Kosuke Yoshihara, Zhubo Wei, Jean C Tien, Roberto Rangel, Kae Hashimoto, Seiji Mabuchi, Kenjiro Sawada, Tadashi Kimura, Neal G Copeland, Nancy A Jenkins
ABSTRACT
Epithelial ovarian cancer (EOC) is a deadly cancer, and its prognosis has not been changed significantly during several decades. To seek new therapeutic targets for EOC, we performed an in vivo dropout screen in human tumor xenografts using a pooled shRNA library targeting thousands of druggable genes. Then, in follow-up studies, we performed a second screen using a genome-wide CRISPR/Cas9 library. These screens identified 10 high-confidence drug targets that included well-known oncogenes such as ERBB2 and RAF1, and novel oncogenes, notably KPNB1, which we investigated further. Genetic and pharmacological inhibition showed that KPNB1 exerts its antitumor effects through multiphase cell cycle arrest and apoptosis induction. Mechanistically, proteomic studies revealed that KPNB1 acts as a master regulator of cell cycle-related proteins, including p21, p27, and APC/C. Clinically, EOC patients with higher expression levels of KPNB1 showed earlier recurrence and worse prognosis than those with lower expression levels of KPNB1. Interestingly, ivermectin, a Food and Drug Administration-approved antiparasitic drug, showed KPNB1-dependent antitumor effects on EOC, serving as an alternative therapeutic toward EOC patients through drug repositioning. Last, we found that the combination of ivermectin and paclitaxel produces a stronger antitumor effect on EOC both in vitro and in vivo than either drug alone. Our studies have thus identified a combinatorial therapy for EOC, in addition to a plethora of potential drug targets.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Nocodazolo, ≥99% (TLC), powder
Sigma-Aldrich
MISSION® esiRNA, targeting human KPNB1