Merck
Decreased WWOX expression promotes angiogenesis in osteosarcoma.
Oncotarget (2017-10-06)
Jia Wen, Zongchao Xu, Jiazhen Li, Yingqiang Zhang, Wenzhe Fan, Yu Wang, Mingjian Lu, Jiaping Li
ABSTRACT
WWOX (WW domain-containing oxidoreductase) is known to be an important tumor suppressor in cancer. In this study, we used samples from 201 osteosarcoma patients to investigate the effects of WWOX on angiogenesis and invasion. WWOX levels were negatively correlated with RUNX2 and VEGF levels, but were not correlated with OPN levels. Among the clinicopathological characteristics examined, WWOX was associated only with response to neoadjuvant chemotherapy, and its expression in osteosarcoma tissues was a predictor of disease-free survival. WWOX promoted apoptosis and inhibited invasion and expression of bcl-2, OPN, RUNX2, and VEGF in osteosarcoma cells
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
MISSION® esiRNA, targeting human WWOX