- Effects of sublethal doses of thiacloprid and its formulation Calypso
Effects of sublethal doses of thiacloprid and its formulation Calypso
The Journal of experimental biology (2017-08-19)
Léa Tison, Sophie Holtz, Amy Adeoye, Önder Kalkan, Nina S Irmisch, Nadja Lehmann, Randolf Menzel
PMID28819056
ABSTRACT
Learning and memory play a central role in the behavior and communication of foraging bees. We have previously shown that chronic uptake of the neonicotinoid thiacloprid affects the behavior of honey bees in the field. Foraging behavior, homing success, navigation performance and social communication were impaired. Thiacloprid collected at a feeding site at low doses accumulates in foragers over time. Here, we applied a laboratory standard procedure (the proboscis-extension response conditioning) in order to assess which processes, acquisition, memory consolidation and/or memory retrieval were compromised after bees were fed either with thiacloprid or the formulation of thiacloprid named Calypso
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto