Merck
  • MiR-361-5p inhibits glycolytic metabolism, proliferation and invasion of breast cancer by targeting FGFR1 and MMP-1.
MiR-361-5p inhibits glycolytic metabolism, proliferation and invasion of breast cancer by targeting FGFR1 and MMP-1.
Journal of experimental & clinical cancer research : CR (2017-11-15)
Fei Ma, Lei Zhang, Li Ma, Yiyun Zhang, Jianguo Zhang, Baoliang Guo
ABSTRACT
MicroRNAs function as key regulators in various human cancers, including breast cancer (BC). MiR-361-5p has been proved to be a tumor suppressor in colorectal cancer and gastric cancer in our previous study. In this study, we aim to find out the function of miR-361-5p in breast cancer progression and elaborate the mechanism that miR-361-5p acts its function in breast cancer. Here we reported that miR-361-5p was down-regulated in breast cancer tissue compared with normal breast tissue and the expression of miR-361-5p was positively associated with prognosis in BC patients. Functional studies showed that overexpression of miR-361-5p suppressed the proliferation, invasion and metastasis of breast cancer cells both in vivo and in vitro. Mechanistically, we found that miR-361-5p inhibited the proliferation of BC cells by suppressing glycolysis. FGFR1, a promoter of glycolysis-related enzyme, was identified as the target of miR-361-5p that promoted glycolysis and repressed oxidative phosphorylation. Furthermore, we demonstrated that miR-361-5p inhibited breast cancer cells invasion and metastasis by targeting MMP-1. An inverse expression pattern was also found between miR-361-5p and FGFR1 or MMP-1 in a cohort of 60 BC tissues. Our results indicate that miR-361-5p inhibits breast cancer cells glycolysis and invasion by respectively repressing FGFR1 and MMP-1, suggesting that miR-361-5p and its targets may serve as therapeutic targets in breast cancer treatment.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
MISSION® esiRNA, targeting human FGFR1 (1)