Merck
MiR-150 alleviates neuropathic pain via inhibiting toll-like receptor 5.
Journal of cellular biochemistry (2017-07-08)
Li-Juan Ji, Jing Shi, Jing-Min Lu, Qiang-Min Huang
ABSTRACT
MicroRNAs (miRNAs) are reported as vital participators in the pathophysiological course of neuropathic pain. However, the underlying mechanisms of the functional roles of miRNAs in neuropathic pain are largely unknown. This study was designed to explore the potential role of miR-150 in regulating the process of neuropathic pain in a rat model established by chronic sciatic nerve injury (CCI). Overexpression of miR-150 greatly alleviated neuropathic pain development and reduced inflammatory cytokine expression, including COX-2, interleukin IL-6, and tumor necrosis factor (TNF)-α in CCI rats. By bioinformatic analysis, 3'-untranslated region (UTR) of Toll-like receptor (TLR5) was predicted to be a target of miR-150. TLR5 commonly serves as an important regulator of inflammation. Overexpression of miR-150 significantly suppressed the expression of TLR5 in vitro and in vivo. Furthermore, upregulation of TLR5 decreased the miR-150 expression and downregulation of TLR5 increased miR-150, respectively. Overexpression of TLR5 significantly reversed the miR-150-induced suppressive effects on neuropathic pain. In conclusion, our current study indicates that miR-150 may inhibit neuropathic pain development of CCI rats through inhibiting TLR5-mediated neuroinflammation. Our findings suggest that miR-150 may provide a novel therapeutic target for neuropathic pain treatment.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
MISSION® esiRNA, targeting human TLR5