Merck
Rapidly Synthesized, Few-Layered Pseudocapacitive SnS
ACS applied materials & interfaces (2017-10-28)
Ranjith Thangavel, Amaresh Samuthira Pandian, Hari Vignesh Ramasamy, Yun-Sung Lee
ABSTRACT
The abundance of sodium resources has recently motivated the investigation of sodium ion batteries (SIBs) as an alternative to commercial lithium ion batteries. However, the low power and low capacity of conventional sodium anodes hinder their practical realization. Although most research has concentrated on the development of high-capacity sodium anodes, anodes with a combination of high power and high capacity have not been widely realized. Herein, we present a simple microwave irradiation technique for obtaining few-layered, ultrathin two-dimensional SnS
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Tin(II) chloride, anhydrous, powder, ≥99.99% trace metals basis