- Lysophosphatidic Acid Receptor 1 Is Important for Intestinal Epithelial Barrier Function and Susceptibility to Colitis.
Lysophosphatidic Acid Receptor 1 Is Important for Intestinal Epithelial Barrier Function and Susceptibility to Colitis.
The American journal of pathology (2017-11-13)
Songbai Lin, Yiran Han, Kayte Jenkin, Sei-Jung Lee, Maiko Sasaki, Jan-Michael Klapproth, Peijian He, C Chris Yun
PMID29128569
ABSTRACT
Intestinal epithelial cells form a barrier that is critical in protecting the host from the hostile luminal environment. Previously, we showed that lysophosphatidic acid (LPA) receptor 1 regulates proliferation of intestinal epithelial cells, such that the absence of LPA1 mitigates the epithelial wound healing process. This study provides evidence that LPA1 is important for the maintenance of epithelial barrier integrity. The epithelial permeability, determined by fluorescently labeled dextran flux and transepithelial resistance, is increased in the intestine of mice with global deletion of Lpar1, Lpar1
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto