Merck
Mortalin inhibitors sensitize K562 leukemia cells to complement-dependent cytotoxicity.
International journal of cancer (2009-09-10)
David Pilzer, Moran Saar, Keizo Koya, Zvi Fishelson
ABSTRACT
Mortalin, the mitochondrial hsp70, is a vital constitutively expressed heat shock protein. Its elevated expression has been correlated with malignant transformation and poor cancer prognosis. Cancer cells exhibit increased resistance to complement-dependent cytotoxicity, partly due to their capacity to eliminate the complement membrane attack complex (MAC) from their cell surface. As we have previously reported, mortalin and the complement membrane attack complexes are released in membrane vesicles from complement attacked cells. As shown here, knock down of mortalin with specific siRNA reduces MAC elimination and enhances cell sensitivity to MAC-induced cell death. Similar results were obtained with MKT-077, a cationic rhodacyanine dye that inhibits mortalin. Treatment of human erythroleukemia K562 and colorectal carcinoma HCT116 cells with MKT-077 sensitizes them to cell death mediated by MAC but not by streptolysin O. Pre-treatment of cells with MKT-077 also reduces the extent of MAC-mortalin vesiculation following a sublytic complement attack. In the presence of MKT-077, the direct binding of mortalin to complement C9, the major MAC component, is inhibited. The tumor suppressor protein p53 is a known mortalin client protein. The effect of MKT-077 on complement-mediated lysis of HCT116 p53(+/+) and p53(-/-) cells was found to be independent on the presence of p53. Our results also demonstrate that recombinant human mortain inhibits complement-mediated hemolysis of rabbit erythrocytes as well as zinc-induced C9 polymerization. We conclude that mortalin supports cancer cell resistance to complement-dependent cytotoxicity and propose consideration of mortalin as a novel target for cancer adjuvant immunotherapy.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Mortalin (47-end), GST tagged human, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution