コンテンツへスキップ
Merck
[Rare cause for severe hypertriglyceridemia - case 9/2013].
Deutsche medizinische Wochenschrift (1946) (2013-11-14)
Sabine Kahl, Michael Roden, Klaus Mörike, Karsten Müssig
要旨
We report on a 48-year-old female patient with recently developed severe hypertriglyceridemia. Medical history was remarkable for breast cancer with breast-preserving surgery and chemoradiotherapy. The patient has been treated with 20 mg tamoxifen per day for three months. Laboratory results showed hypertriglyeridemia, hypercholesterolemia and lowered HDL-cholesterol. Findings were consistent with a drug-induced hypertriglyceridemia caused by anti-estrogenic therapy with tamoxifen. After consulting the patient's gynaecologist, we discontinued tamoxifen treatment. Thereupon, triglyceride levels fell consistently. There were no signs of pancreatitis, serum amylase and lipase were in the normal range. Patients with pre-diagnosed metabolic disorders, especially dyslipidemia and type 2 diabetes, should undergo regular controls of serum triglycerides during tamoxifen treatment. Also, one should keep in mind that a subacute, severe rise in serum triglyceride levels may be caused, in rare cases, by tamoxifen treatment.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
タモキシフェン, ≥99%
Sigma-Aldrich
タモキシフェン クエン酸塩, ≥99%
タモキシフェン クエン酸塩, European Pharmacopoeia (EP) Reference Standard
タモキシフェン クエン酸塩, European Pharmacopoeia (EP) Reference Standard