제품 이름
Simvastatin, Sodium Salt - Calbiochem,
양식
(Beige solid)
Quality Level
저장 조건
OK to freeze
solubility
soluble (DMSO (100 mg/ml), Ethanol (25 mg/ml), or H2O (5 mg/ml))
저장 온도
−20°C
일반 설명
Carboxylate form of Simvastatin (Cat. No. 567020) that is active in whole cells and in cell-free assays. A lipophilic 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase inhibitor that blocks Ras function through inhibition of farnesylation. Also inhibits glucose-induced Ca2+-signaling and insulin secretion by blocking L-type Ca2+ channels in rat islet β-cells. Inhibits cell proliferation of human smooth muscle cells. Note: This product is supplied in an active form; no treatment with NaOH in ethanol is required for its activation.