제품 이름
Omnifit® EZ Benchmark Columns with 1 Adjustable and 1 fixed Endpiece, I.D. × L 25 mm × 500 mm
manufacturer/tradename
Omnifit 006EZ-25-50-AF
technique(s)
LPLC: suitable
I.D. × L
25 mm × 500 mm
Quality Level
유사한 제품을 찾으십니까? 방문 제품 비교 안내
General description
Omnifit® Labware complete columns are supplied assembled and ready for use, with tubing and fittings to connect the column into an existing system. Two configurations are available:
- Non-adjustable with two fixed endpieces (-FF), a cost-effective option when height adjustment is not important.
- One fixed endpiece as a bed support and one adjustable endpiece (plunger) to adjust the bed height (-AF). The adjustable endpiece is designed to give the user fine control over the movement of the endpiece, minimizing the risk of disturbance to the packed bed and giving 80mm of height adjustment.
Legal Information
Omnifit is a registered trademark of Diba Industries