Merck
  • Human serum potentiates the expression of genes associated with antifungal drug resistance in C. albicans biofilms on central venous catheters.
Human serum potentiates the expression of genes associated with antifungal drug resistance in C. albicans biofilms on central venous catheters.
Mycopathologia (2014-12-18)
L P Samaranayake, S Anil, M Hashem, S Vellappally, B P K Cheung
RESUMEN
Candida albicans is a major agent of fungaemias and frequently causes systemic disease through seeded, blood stream dissemination. These infections, particularly common in hospitalized patients with central venous catheters (CVCs), appear to persevere due to biofilm reservoirs of the yeast that tend to develop on the device. Although it is known that candidal biofilms are intrinsically resistant to antifungals compared with their planktonic counterparts, there is a paucity of data on the expression of antifungal drug resistance genes (DRGs) in candidal biofilms in CVC reservoirs. Furthermore, notwithstanding the fact that CVCs are constantly bathed in human serum, there are no studies on the effect of the latter on the DRG expression in candidal biofilms. Hence, we developed in vitro biofilms of three different C. albicans strains on silicone CVC discs immersed in human serum and evaluated the temporal expression of nine antifungal DRGs. In an attempt to evaluate the effect of hyphal elements on DRG expression, we incorporated a hyphal mutant (HM) and its wild-type (WT) counterpart, as well as a fresh clinical isolate in the studies. Human serum significantly up-regulated DRG transcripts in Candida biofilms on CVCs, at different stages of biofilm growth, while the WT strain over-expressed more DRGs than the HM strain. Here, we report, for the first time, that both human serum and the hyphal elements of the yeast have a profound modulatory effect on DRG expression in C. albicans biofilms on CVCs.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Alcohol etílico puro 200, Molecular Biology
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Alcohol etílico puro, 200 proof, ACS reagent, ≥99.5%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Alcohol etílico puro, 200 proof
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Alcohol etílico puro, 200 proof, meets USP testing specifications
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
D-(+)-Glucosa, ≥99.5% (GC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
D-(+)-Glucosa, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Alcohol etílico puro, 200 proof, anhydrous, ≥99.5%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Alcohol etílico puro 190, for molecular biology
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Dextrosa, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Alcohol etílico puro, 190 proof, ACS spectrophotometric grade, 95.0%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
D-(+)-Glucosa, ≥99.5% (GC), BioXtra
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Etanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Supelco
D-(+)-Glucosa, Pharmaceutical Secondary Standard; Certified Reference Material
En este momento no podemos mostrarle ni los precios ni la disponibilidad
USP
D-(+)-Glucosa, United States Pharmacopeia (USP) Reference Standard
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
D-(+)-Glucosa, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Supelco
Etanol, Pharmaceutical Secondary Standard; Certified Reference Material
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
D-(+)-Glucosa, ACS reagent
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Alcohol etílico puro, 190 proof, meets USP testing specifications
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Supelco
D-(+)-Glucosa, analytical standard
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Menadione, crystalline
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Supelco
Etanol, standard for GC
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Menadione, meets USP testing specifications
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
D-(+)-Glucosa, suitable for mouse embryo cell culture, ≥99.5% (GC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Supelco
Ethanol solution, 10 % (v/v) in H2O, analytical standard
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Supelco
Menadione (K3), analytical standard
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
D-(+)-Glucosa, Hybri-Max, powder, BioReagent, suitable for hybridoma
En este momento no podemos mostrarle ni los precios ni la disponibilidad
USP
Etanol, United States Pharmacopeia (USP) Reference Standard
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
D-(+)-Glucosa, tested according to Ph. Eur.
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Supelco
Menadione, Pharmaceutical Secondary Standard; Certified Reference Material
En este momento no podemos mostrarle ni los precios ni la disponibilidad