Skip to Content
Merck

BDPH9BCD264F

Bide Pharmatech

4-Chloro-6-(trifluoromethyl)quinazoline

Synonym(s):

4-chloro-6-(trifluoromethyl)quinazoline

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H4N2Cl1F3
CAS Number:
Molecular Weight:
232.59
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H4ClF3N2/c10-8-6-3-5(9(11,12)13)1-2-7(6)14-4-15-8/h1-4H

Inchi Key

OJNBXNWWYXBHGZ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

White to Pale-yellow to Yellow-brown Solid



Still not finding the right product?


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library