Skip to Content

BDPH9BCDA71B

Bide Pharmatech

4-Amino-2-fluoro-N-methylbenzamide

Synonym(s):

4-amino-2-fluoro-N-methylbenzamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H9O1N2F1
CAS Number:
Molecular Weight:
168.17
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H9FN2O/c1-11-8(12)6-3-2-5(10)4-7(6)9/h2-4H,10H2,1H3,(H,11,12)

Inchi Key

XOKAXPQJUODMSH-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Pale-yellow to Yellow-brown to Brown Solid



Still not finding the right product?

Explore all of our products under 4-Amino-2-fluoro-N-methylbenzamide


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library