Merck
  • Pyrrolidine based chiral organocatalyst for efficient asymmetric Michael addition of cyclic ketones to β-nitrostyrenes.
Pyrrolidine based chiral organocatalyst for efficient asymmetric Michael addition of cyclic ketones to β-nitrostyrenes.
Bioorganic & medicinal chemistry letters (2012-06-08)
Kamal Nain Singh, Paramjit Singh, Pushpinder Singh, Nand Lal, Sandeep Kumar Sharma
ABSTRACT
An efficient asymmetric Michael addition of cyclic ketones to β-nitrostyrenes using secondary diamine as an organocatalyst derived from l-proline and (R)-α-methylbenzyl amine has been described. This pyrrolidine based catalyst 1 was found to be very effective to synthesize various γ-nitrocarbonyl compounds in good yield (up to 81%) with excellent stereoselectivity (up to >99:1 dr and >99% ee).
Sigma-Aldrich
trans-β-Nitrostyrene, 99%
Pricing and availability is not currently available.