Skip to Content
Merck

Skip To

V697230

[1,1′-Bis(diphenylphosphino)ferrocene]dichloropalladium(II)

powder, Vetec™

Synonym(s):

Pd(dppf)Cl2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
(C17H14P)2Fe · PdCl2
CAS Number:
Molecular Weight:
731.70
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

[1,1′-Bis(diphenylphosphino)ferrocene]dichloropalladium(II), Vetec™

Quality Segment

form

powder

reaction suitability

core: palladium, reaction type: Buchwald-Hartwig Cross Coupling Reaction, reaction type: Cross Couplings, reaction type: Heck Reaction, reaction type: Hiyama Coupling, reaction type: Negishi Coupling, reaction type: Sonogashira Coupling, reaction type: Stille Coupling, reaction type: Suzuki-Miyaura Coupling

mp

266-283 °C (lit.)

storage temp.

room temp

SMILES string

[Fe].Cl[Pd]Cl.[CH]1[CH][CH][C]([CH]1)P(c2ccccc2)c3ccccc3.[CH]4[CH][CH][C]([CH]4)P(c5ccccc5)c6ccccc6

InChI

1S/2C17H14P.2ClH.Fe.Pd/c2*1-3-9-15(10-4-1)18(17-13-7-8-14-17)16-11-5-2-6-12-16;;;;/h2*1-14H;2*1H;;/q;;;;;+2/p-2

InChI key

JCWIWBWXCVGEAN-UHFFFAOYSA-L

Application

Catalyst for C-C and C-N coupling reaction.
Pd(dppf)Cl2 may be used as an effective palladium catalyst in the following reactions:

  • Cross-coupling of sec-alkyl and n-alkyl Grignard reagents with high yield and selectivity.
  • Suzuki coupling of aryl boronic esters with [11C]methyl iodide to form functionalized [11C]toluene derivatives
  • Kumada cross-coupling of 1,3,5-tribromobenzene with Grignard reagents to form star-shaped oligothiophenes
For small scale and high throughput uses, product is also available as ChemBeads (919780)

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?


Storage Class

10 - Combustible liquids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library