Przejdź do
BR782800
BRAND® insert strips for 24 x 6-well BRANDplates®
PC membrane, pore size 0.4 μm
Synonim(y):
BRANDplates®, Cell culture, Insert Strips, Insert System, Inserts, Tissue culture
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
Informacje o tej pozycji
NACRES:
NB.22
UNSPSC Code:
41122107
eCl@ss:
32035490
Material:
PC membrane
Pore size:
0.4 μm
Sterility:
sterile
Binding type:
non-treated surface
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomóc
material
PC membrane
description
cellGrade™ plus
sterility
sterile
packaging
pack of 12 ea (strips of 4 inserts, individually wrapped)
manufacturer/tradename
BRAND 782800
pore size
0.4 μm
binding type
non-treated surface
General description
Innovative cell culture plates and inserts for manual and automatic cell and tissue culture. The BRANDplates® Insert System, developed in conjunction with the Fraunhofer company, offers an innovative expansion to BRAND′s existing cell culture product range. It consists of two different cell culture plates and two designs of cell culture inserts that can be combined in different configurations.Insert specification:4x insert strips, can be divided into up to 4 individual inserts - hanging position in wellHanging inserts – distance between the well base and insert membrane is 4 mmIndividually packaged strips inserted in thermoformed beakers with 6 strips each in the 6-well cell culture platecellGrade™ plus (hydrophilic cell culture-) surface.Smooth wall inserts: For standard applications like secretion studies, co-cultivations, chemotaxis tests, etc.PS cellGrade™ plus surface, sterile, strip of 4 inserts (divisible).BIO-CERT® CELL CULTURE QUALITY: Free of human DNA, RNase, DNase, Pyrogens and non cytotoxic
Legal Information
BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG
cellGrade is a trademark of BRAND GMBH + CO KG
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
BR782801BR782861BR782821
description
PC membrane, pore size 0.4 μm
description
Inlet Opening System, PC membrane, size 0.4 μm
description
Inlet Opening System, PC membrane, pore size 8 μm
description
Inlet Opening System, PC membrane, pore size 1 μm
sterility
sterile
sterility
sterile
sterility
sterile
sterility
sterile
binding type
non-treated surface
binding type
non-treated surface
binding type
non-treated surface
binding type
-
pore size
0.4 μm
pore size
0.4 μm
pore size
8 μm
pore size
1 μm
manufacturer/tradename
BRAND 782800
manufacturer/tradename
BRAND 782801
manufacturer/tradename
BRAND 782861
manufacturer/tradename
BRAND 782821
material
PC membrane
material
PC membrane
material
PC membrane
material
PC membrane
packaging
pack of 12 ea (strips of 4 inserts, individually wrapped)
packaging
pack of 12 ea (strips of 4 inserts, individually wrapped), pkg of (individually wrapped)
packaging
pack of 12 ea (strips of 4 inserts, individually wrapped)
packaging
pack of 12 ea (strips of 4 inserts, individually wrapped)
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)