Informacje o tej pozycji
NACRES:
NB.51
UNSPSC Code:
41121800
eCl@ss:
32110208
Pomoc techniczna
Pozwól nam pomócjoint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C204250
jacket L
250 mm
General description
Synthware™ Liebig condenser features a 250 mm jacketed length for superior cooling efficiency. With standard taper outer joint at the top (24/40) and a lower inner drip tip joint. Made from durable borosilicate glass.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Still not finding the right product?
Explore all of our products under Synthware™ Liebig condenser with 8 mm OD hose connections
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?