Zmień widok
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
NACRES:
NB.51
UNSPSC Code:
41121800
Manufacturer/tradename:
Synthware C414250
Capacity:
250 mL
Material:
colorless , glass
Sterility:
non-sterile
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomócmaterial
colorless , glass
joint
joint: ST/NS24/40
sterility
non-sterile
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C414250
capacity
250 mL
General description
Reservoir has a top 24/40 or 45/50 Rodaviss outer joint for use with flow controller, and the listed size lower inner joint. Supplied complete with open top cap, Viton® O-ring and loosening ring.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description capacity 250 mL, joint: ST/NS 24/40 | description capacity 500 mL, joint: ST/NS 24/40 | description capacity 250 mL, joint: ST/NS 24/40 | description joint: ST/NS 24/40 |
| capacity 250 mL | capacity 500 mL | capacity 250 mL | capacity - |
| sterility non-sterile | sterility non-sterile | sterility non-sterile | sterility - |
| manufacturer/tradename Synthware C414250 | manufacturer/tradename Synthware C414500 | manufacturer/tradename Synthware C182402 | manufacturer/tradename C432440 |
| material glass , colorless | material colorless , glass | material colorless , glass | material - |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pack of 1 ea |
| joint joint: ST/NS24/40 | joint joint: ST/NS24/40 | joint joint: ST/NS24/40 | joint joint: ST/NS24/40 |
Still not finding the right product?
Explore all of our products under Synthware™ Chromatography reservoir with Rodaviss joint
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?