Przejdź do
P-210
Phenmetrazine hydrochloride solution
100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Synonim(y):
Phenmetrazine hydrochloride, 3-Methyl-2-phenylmorpholine hydrochloride
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
Informacje o tej pozycji
Wzór empiryczny (zapis Hilla):
C11H15NO · HCl
Numer CAS:
Masa cząsteczkowa:
213.70
UNSPSC Code:
41116107
eCl@ss:
32150313
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomóc
grade
certified reference material
Quality Segment
feature
Snap-N-Spike®/Snap-N-Shoot®
packaging
ampule of 1 mL
manufacturer/tradename
Cerilliant®
drug control
Pszichotróp anyag / Psychotropic Substance (Hungary), 78/2022. (XII. 28.) BM rendelet, kontrollierte Droge in Deutschland
concentration
100 μg/mL in methanol (as free base)
format
single component solution
storage temp.
-10 to -25°C
InChI
1S/C11H15NO.ClH/c1-9-11(13-8-7-12-9)10-5-3-2-4-6-10;/h2-6,9,11-12H,7-8H2,1H3;1H
InChI key
VJNXVAVKCZJOFQ-UHFFFAOYSA-N
General description
Phenmetrazine, marketed under the names as Preludin, is a sympathomimetic drug that acts as a stimulant in the central nervous system primarily used as appetite suppressant. This Certified Spiking Solution® is applicable for use in clinical toxicology, forensic analysis, urine drug testing, or pharmaceutical research applications by LC-MS/MS or GC/MS.
Legal Information
German
Dieses Produkt fällt unter das Betäubungsmittelgesetz (BtMG). Für eine Bestellung dieses Produktes ist eine Erlaubnis nach § 3 BtMG zwingend erforderlich, es sei denn, es greift eine Ausnahme von der Erlaubnispflicht nach § 4 oder § 26 BtMG.
English
This product is subject to the German Narcotics Act. A permit under Section 3 of the German Narcotics Act is mandatory for ordering this product unless an exemption from the permit requirement under Section 4 or Section 26 of the German Narcotics Act applies.
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
D159
description
100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
description
1 mg/mL in acetonitrile, certified reference material, ampule of 1 mL, Cerilliant®
format
single component solution
format
single component solution
grade
certified reference material
grade
certified reference material
drug control
Pszichotróp anyag / Psychotropic Substance (Hungary), 78/2022. (XII. 28.) BM rendelet, kontrollierte Droge in Deutschland
drug control
kontrollierte Droge in Deutschland, Pszichotróp anyag / Psychotropic Substance (Hungary), 78/2022. (XII. 28.) BM rendelet
Quality Level
300
Quality Level
300
concentration
100 μg/mL in methanol (as free base)
concentration
1 mg/mL in acetonitrile
storage temp.
-10 to -25°C
storage temp.
−20°C
Słowo ostrzegawcze
Danger
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
Narządy docelowe
Eyes,Central nervous system
Klasa składowania
3 - Flammable liquids
WGK
WGK 2
Temperatura zapłonu (°F)
49.5 °F - closed cup
Temperatura zapłonu (°C)
9.7 °C - closed cup
Wykazy regulacyjne
Wykazy regulacyjne dotyczą głównie produktów chemicznych. Można w nich podawać ograniczoną liczbę informacji na temat produktów niechemicznych. Brak wpisu oznacza, że żaden ze składników nie znajduje się w wykazie. Użytkownik odpowiada za zagwarantowanie bezpiecznego i zgodnego z prawem stosowania produktu.
67-56-1
CAS No.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)