Zaloguj się, aby wyświetlić ceny organizacyjne i kontraktowe.
Wybierz wielkość
Zmień widok
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
UNSPSC Code:
12161503
NACRES:
NA.21
Technique(s):
single molecule counting: suitable
Compatibility:
for use with FemtoQuest™ and SMCxPRO® instruments
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Potrzebujesz pomocy? Nasz zespół doświadczonych naukowców chętnie Ci pomoże.
Pozwól nam pomócQuality Segment
description
Used to determine assay buffer conditions needed for optimal SMC® immunoassay performance.
manufacturer/tradename
SMC®
packaging
pkg of 96 well assay
technique(s)
single molecule counting: suitable
compatibility
for use with FemtoQuest™ and SMCxPRO® instruments
shipped in
wet ice
storage temp.
2-8°C
Features and Benefits
- Increased Efficiency: Accelerate assay development lead times by rapidly & systematically screening a range of buffer formulations.
- Optimized Performance: Reduce background noise and non-specific binding while improving assay stringency.
- Ready-to-use Format: Seamlessly integrates into the assay development workflow of the SMC® Bead-Based Assay Development Kit (03-0222-00).
Other Notes
This kit should be used in conjunction with the SMC® Bead-Based Assay Development Kit (03-0222-00).
Buffers contain passive blockers to prevent non-specific binding. Active blockers appropriate for the assay being developed may need to be added and optimized to further reduce non-specific binding and enhance specificity and/or sensitivity.
Buffers contain passive blockers to prevent non-specific binding. Active blockers appropriate for the assay being developed may need to be added and optimized to further reduce non-specific binding and enhance specificity and/or sensitivity.
Legal Information
FEMTOQUEST is a trademark of Merck KGaA, Darmstadt, Germany
SMC is a registered trademark of Singulex
SMCxPRO is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Ta strona może zawierać tekst przetłumaczony maszynowo.
Kody zagrożeń
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1
Klasa składowania
10 - Combustible liquids
Co się dzieje?
WGK 3
Słowo ostrzegawcze
Danger
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?
Dokumenty związane z niedawno zakupionymi produktami zostały zamieszczone w Bibliotece dokumentów.

