Cheap, effective, easy to clean. They seem to keep their shape somewhat after washing, even if bent out of shape while transferring.
Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
5 ea | Dostępne do wysyłki DZISIAJzKuehne + Nagel Sp. z o.o. | 75,30 zł |
Informacje o tej pozycji
75,30 zł
Dostępne do wysyłki DZISIAJSzczegóły
packaging
pkg of 5 packs
manufacturer/tradename
Millipore
technique(s)
electrophoresis: suitable, immunoblotting: suitable, western blot: suitable
compatibility
for use with mPAGE® Mini Wet Transfer System
General description
The mPAGE® Mini Wet Transfer System (MWTS) accommodates two gel holder cassettes for electrophoretic transfer of proteins from mini gels to PVDF or nitrocellulose blotting membrane using mPAGE® Transfer Buffer or a compatible transfer buffer.
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description replacement part for mPAGE® Mini Wet Transfer System | description for use with mPAGE® Mini Wet Transfer System | description for use with mPAGE® Gel Caster and mPAGE® Mini Gel Tank | description replacement part for mPAGE® Gel Caster |
| compatibility for use with mPAGE® Mini Wet Transfer System | compatibility for use with mPAGE® Mini Wet Transfer System | compatibility for use with mPAGE® Gel Caster | compatibility for use with
|
| packaging pkg of 5 packs | packaging pkg of 1 pack | packaging pkg of 5 packs | packaging pkg of 5 packs |
| manufacturer/tradename Millipore | manufacturer/tradename Millipore | manufacturer/tradename Millipore | manufacturer/tradename Millipore |
| technique(s) immunoblotting: suitable, electrophoresis: suitable | technique(s) electrophoresis: suitable, western blot: suitable, immunoblotting: suitable | technique(s) electrophoresis: suitable, SDS-PAGE: suitable | technique(s) electrophoresis: suitable, SDS-PAGE: suitable |
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?