Przejdź do
GREL5
MPAGE®® Gel Releasers
for use with mPAGE® Gel Caster and mPAGE® Mini Gel Tank
1 of 1 reviewers received a sample product or took part in a promotion
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
5 ea
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
90,70 zł
Informacje o tej pozycji
UNSPSC Code:
41105305
NACRES:
NC.01
eCl@ss:
32039090
90,70 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
​
Pomoc techniczna
Pozwól nam pomóc
Nazwa produktu
mPAGE® Gel Releasers, 5pk, for use with mPAGE® Gel Caster and mPAGE® Mini Gel Tank
packaging
pkg of 5 packs
manufacturer/tradename
Millipore
technique(s)
SDS-PAGE: suitable, electrophoresis: suitable
compatibility
for use with mPAGE® Gel Caster
General description
Use mPAGE® Gel Releasers to remove polyacrylamide gels cast with the mPAGE® Gel Caster.
The mPAGE® Gel Caster offers a leak-free solution for handcasting up to 2 mini protein gels. Gels cast
are 8 cm x 10 cm with a gel thickness of 1.0 mm, 0.75 mm, or 1.5 mm, along
with 10 or 15-well options for sample loading. The mPAGE® Gel Caster has been
designed to simplify hand casting gels and to eliminate leakage during the casting
process. The gel caster is available as a standalone device (GCR2) or as a gel casting device kit for casting mini gels gels with 0.75 mm, 1.0 mm and 1.5 mm thickness (MGCK-75M, MGCK-10M, MGCK-15M).
Legal Information
MPAGE is a registered trademark of Merck KGaA, Darmstadt, Germany
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
GCR2GCBASEMSPA75
description
for use with mPAGE® Gel Caster and mPAGE® Mini Gel Tank
description
2 pack, device for hand casting mini gels, does not include plates or combs
description
replacement part for mPAGE® Gel Caster
description
for use with mPAGE® Gel Caster
compatibility
for use with mPAGE® Gel Caster
compatibility
for use with mPAGE®
  • TurboMix Bis-Tris Gel Casting Kit
  • Combs
  • Mini Spacer Plates
  • Short Plates
  • Mini Gel Tank
  • Mini Wet Transfer System
compatibility
for use with
  • mPAGE® Combs 10 wells (0.75 mm, 1.0 mm or 1.5 mm)
  • mPAGE® Combs 15 wells (0.75 mm, 1.0 mm or 1.5 mm)
  • mPAGE® Mini Spacer Plates (0.75 mm, 1.0 mm or 1.5 mm)
  • mPAGE® Mini Short Plates
compatibility
for casting gels with the mPAGE® Gel Caster components:
  • mPAGE® Gel Caster Base
  • mPAGE® Gel Caster Frame
  • mPAGE® Gel Caster Sealing Gaskets
compatible with mPAGE® Combs:
  • 0.75 mm, 10 wells
  • 0.75 mm, 15 wells
packaging
pkg of 5 packs
packaging
pack of 2
packaging
pkg of 1 pack
packaging
pkg of 5 packs
manufacturer/tradename
Millipore
manufacturer/tradename
Millipore
manufacturer/tradename
Millipore
manufacturer/tradename
Millipore
technique(s)
SDS-PAGE: suitable
technique(s)
SDS-PAGE: suitable, electrophoresis: suitable
technique(s)
SDS-PAGE: suitable, electrophoresis: suitable
technique(s)
electrophoresis: suitable, SDS-PAGE: suitable
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)
Active Filters
  1. Toronto, ON
    • Reviews 4
    • Vote 1
    5 out of 5 stars.
    Simple, handy
    They're pieces of plastic that work well for what they do. A bit too sharp to transfer the gel for transfer, but we can use a spatula for that.
    Helpful?