Informacje o tej pozycji
NACRES:
SB.51
UNSPSC Code:
41104200
eCl@ss:
32039090
Pomoc techniczna
Pozwól nam pomócAC/DC input
230 V, 50-60 Hz
packaging
pkg of 1 unit
compatibility
for use with tanks
General description
Recirculation Pump is a water purification system accessory. The pump recirculates stored water to maintain pure water quality.
For use with 30, 60, and 100 L polyethylene (PE) reservoirs to protect stored water quality.
For use with 30, 60, and 100 L polyethylene (PE) reservoirs to protect stored water quality.
Still not finding the right product?
Explore all of our products under Recirculation Pump
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?