Przejdź do
Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 2140,00 zł |
Informacje o tej pozycji
2140,00 zł
grade
certified reference material
agency
traceable to (National Research Council of Canada (NRC))
manufacturer/tradename
National Research Council of Canada (NRC)
application(s)
environmental
format
matrix material
General description
Analysis Note
Other Notes
Trace Elements: Arsenic (As), Boron (B), Cadmium (Cd), Cerium (Ce), Chromium (Cr), Cobalt (Co), Copper (Cu), Dimethylarsenic (as As), Dysprosium (Dy), Erbium (Er), Europium (Eu), Gadolinium (Gd), Holmium (Ho), Inorganic Arsenic, Iron (Fe), Lanthanum (La), Lead (Pb), Lutetium (Lu), Manganese (Mn), Molybdenum (Mo), Monomethylarsenic (as As), Neodymium (Nd), Nickel (Ni), Praseodymium (Pr), Samarium (Sm), Terbium (Tb), Thulium (Tm), Trimethylarsenic (as As), Uranium (U), Vanadium (V), Ytterbium (Yb), Zinc (Zn)
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description certified reference material | description certified reference material | description certified reference material | description certified reference material |
| application(s) environmental | application(s) food and beverages | application(s) cosmetics | application(s) food and beverages |
| format matrix material | format matrix material | format matrix material | format matrix material |
| agency traceable to (National Research Council of Canada (NRC)) | agency (National Research Council of Canada (NRC)) | agency - | agency traceable to (National Research Council of Canada (NRC)) |
| grade certified reference material | grade certified reference material | grade certified reference material | grade certified reference material |
| manufacturer/tradename National Research Council of Canada (NRC) | manufacturer/tradename National Research Council of Canada (NRC) | manufacturer/tradename National Research Council of Canada (NRC) | manufacturer/tradename National Research Council of Canada (NRC) |
Klasa składowania
10 - Combustible liquids
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?
Dokumenty związane z niedawno zakupionymi produktami zostały zamieszczone w Bibliotece dokumentów.

