Przejdź do zawartości
Merck

Przejdź do

CASS-7

Nearshore Seawater Certified Reference Material for Trace Metals and Other Constituents

certified reference material

Synonim(y):

reference material

Zaloguj się, aby wyświetlić ceny organizacyjne i kontraktowe.

Wybierz wielkość

Zmień widok
Rozmiar/SKUDostępnośćCena netto

Informacje o tej pozycji

UNSPSC Code:
41116107
eCl@ss:
25140904
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Potrzebujesz pomocy? Nasz zespół doświadczonych naukowców chętnie Ci pomoże.
Pozwól nam pomóc


grade

certified reference material

agency

(National Research Council of Canada (NRC))

manufacturer/tradename

National Research Council of Canada (NRC)

application(s)

food and beverages

format

matrix material

General description

CASS-7 is a nearshore seawater Certified Reference Material (CRM) prepared at the National Research Council of Canada (NRC) with information on trace element content and other constituents. A unit of CASS-7 consists of approximately 200 mL of seawater. This material is intended for use in the method development, validation, and quality control for the analysis of trace and matrix constituents in seawater matrices. https://doi.org/10.4224/crm.2025.cass-7

Features and Benefits

NRC Canada′s certified reference materials (CRMs) are traceable to the International System of Units, and conform to the requirements of ISO/IEC 17025 and ISO 17034.For specific values, as well as additional information regarding stability and storage information, please view the Certificate of Analysis.With NRC Canada′s CRMs, you gain access to products backed by extensive research and expertise, fostering trust and consistency in your results. Elevate your laboratory′s accuracy and credibility with NRC Canada′s certified reference materials.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  
Ta strona może zawierać tekst przetłumaczony maszynowo.

Porównaj podobne pozycje

Wyświetl pełne porównanie

Pokaż różnice

1 of 1

Ta pozycja
NASS7MOOS4AQUA1
description

certified reference material

description

certified reference material

description

certified reference material

description

certified reference material

application(s)

food and beverages

application(s)

environmental

application(s)

food and beverages

application(s)

cosmetics
food and beverages

format

matrix material

format

matrix material

format

matrix material

format

matrix material

agency

(National Research Council of Canada (NRC))

agency

traceable to (National Research Council of Canada (NRC))

agency

(National Research Council of Canada (NRC))

agency

-

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

manufacturer/tradename

National Research Council of Canada (NRC)

manufacturer/tradename

National Research Council of Canada (NRC)

manufacturer/tradename

National Research Council of Canada (NRC)

manufacturer/tradename

National Research Council of Canada (NRC)


Still not finding the right product?


Klasa składowania

12 - Non Combustible Liquids

Temperatura zapłonu (°F)

Not applicable

Temperatura zapłonu (°C)

Not applicable


Wykazy regulacyjne

Wykazy regulacyjne dotyczą głównie produktów chemicznych. Można w nich podawać ograniczoną liczbę informacji na temat produktów niechemicznych. Brak wpisu oznacza, że żaden ze składników nie znajduje się w wykazie. Użytkownik odpowiada za zagwarantowanie bezpiecznego i zgodnego z prawem stosowania produktu.

7697-37-2

CAS No.


Wybierz jedną z najnowszych wersji:

Certyfikaty analizy (CoA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumenty section.

Proszę o kontakt, jeśli potrzebna jest pomoc Obsługa Klienta

Masz już ten produkt?

Dokumenty związane z niedawno zakupionymi produktami zostały zamieszczone w Bibliotece dokumentów.

Odwiedź Bibliotekę dokumentów