Przejdź do
BUA481F2-EP
Buckets for Eppendorf® A-4-81 Rotor
MTP/Flex bucket, pack of 2
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
Informacje o tej pozycji
UNSPSC Code:
41103910
eCl@ss:
32110390
Manufacturer/tradename:
Eppendorf® 022638866 (NA), Eppendorf® 5810742005 (EU)
Feature:
Holds x flex bucket, MTP/Flex bucket
Produkt BUA481F2-EP nie jest obecnie dostępny w sprzedaży w Twoim kraju Skontaktuj się z zespołem ds. pomocy technicznej
Pomoc techniczna
Pozwól nam pomóc
feature
Holds x flex bucket, MTP/Flex bucket
packaging
pack of 2
manufacturer/tradename
Eppendorf® 022638866 (NA), Eppendorf® 5810742005 (EU)
technique(s)
centrifugation: suitable
General description
MTP/Flex buckets are designed for use with Eppendorf® A-4-81 rotor. These buckets have flat bottoms with rubber mats. MTP/Flex buckets supplied as a set of 2 can accommodate 4 microplates(MTP), 2 cell culture plates, or 1 deep well plate(DWP) per bucket.
Application
MTP/Flex bucket is suitable for A-4-81 rotor for 5910 Ri centrifuge.
Features and Benefits
  • Designed for Eppendorf® A-4-81 and S-4x500 rotors
  • Ideal for use with IsoRack and cell culture flask adapters as well as microplates(MTP), and deep well plates (DWP)
  • MTP/Flex buckets with a maximum speed of 4,000 RPM and a maximum RCF of 2,900 x g are ideal for high-speed centrifugation
  • MTP/Flex buckets have a 163 mm centrifugation radius
Legal Information
Eppendorf is a registered trademark of Eppendorf SE
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)