Przejdź do
BUA481F4-EP
Buckets for Eppendorf® A-4-81 Rotor
MTP/Flex bucket, pack of 4
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
Informacje o tej pozycji
UNSPSC Code:
41103910
eCl@ss:
32110390
Manufacturer/tradename:
Eppendorf® 022638840 (NA), Eppendorf® 5810741009 (EU)
Feature:
Holds x flex bucket, MTP/Flex bucket
Produkt BUA481F4-EP nie jest obecnie dostępny w sprzedaży w Twoim kraju Skontaktuj się z zespołem ds. pomocy technicznej
Pomoc techniczna
Pozwól nam pomóc
feature
Holds x flex bucket, MTP/Flex bucket
packaging
pack of 4
manufacturer/tradename
Eppendorf® 022638840 (NA), Eppendorf® 5810741009 (EU)
technique(s)
centrifugation: suitable
General description
Eppendorf is a well-known company in the field of life sciences. They specialize in creating and selling various tools, materials, and services used in laboratories around the world. Their product lineup includes pipettes, automated pipetting systems, dispensers, centrifuges, mixers, spectrometers, and DNA amplification tools. Eppendorf products are widely used in both academic and commercial research labs. Centrifuges and microcentrifuges find applications in various fields such as microbiology, tissue culture, molecular biology, PCR, genomics, clinical research, drug discovery, and proteomics. We provide a range of centrifuges, rotors, buckets, and adapters designed to meet your specific processing requirements. This bucket can be used for rotors A-4-81, S-4x500. It is suitable for use with IsoRack and cell culture flask adapters as well as MTP and DWP.
Legal Information
Eppendorf is a registered trademark of Eppendorf SE
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)