Przejdź do
Zaloguj się, aby wyświetlić ceny organizacyjne i kontraktowe.
Wybierz wielkość
Zmień widok
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
Biological source:
human (iPSC line)
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Potrzebujesz pomocy? Nasz zespół doświadczonych naukowców chętnie Ci pomoże.
Pozwól nam pomócbiological source
human (iPSC line)
form
frozen liquid
packaging
vial of 1 (contains ≥1 million cells)
technique(s)
cell culture | mammalian: suitable
shipped in
liquid nitrogen
storage temp.
-140 to -196°C
General description
Human iPSC-Derived Midbrain Dopaminergic Neurons. Required supplements for the final cell differentiation and maturation (sold separately): BX-2200-100uL. Dopaminergic neurons are a subtype of neurons that release dopamine as their primary neurotransmitter. Important for various neurological functions including motor coordination and executive function, dopaminergic neurons are a critical part of the central nervous system (CNS). Dopaminergic neurons can be found in the midbrain, and have been a crucial model for the study of regenerative medicine and the development of novel therapeutics for and understand of Parkinson’s disease, Huntington’s disease, ADHD, addiction, and epilepsy. Primary neurotransmitter released: dopamine.
Application
Function: Dopaminergic neurons actively release and uptake dopamine, reflecting their functional maturity. They display both spontaneous and stimulus-evoked electrical activity, making them ideal for electrophysiological applications. Importantly, they are validated for use with microelectrode array (MEA) platforms, allowing researchers to track neuronal network behavior and evaluate how different compounds influence firing patterns.
Features and Benefits
Marker Expression: Fully differentiated Midbrain Dopaminergic Neurons consist of cells derived from human iPSCs that have been induced to neuroepithelial cells, patterned to midbrain floorplate progenitors and differentiated to midbrain dopaminergic neurons. The resulting cell populations contain 50% TH+ cells (a marker for dopaminergic cells).
Ta strona może zawierać tekst przetłumaczony maszynowo.
Słowo ostrzegawcze
Danger
Kody zagrożeń
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Repr. 2 - Skin Sens. 1
Klasa składowania
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Temperatura zapłonu (°F)
Not applicable
Temperatura zapłonu (°C)
Not applicable
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?
Dokumenty związane z niedawno zakupionymi produktami zostały zamieszczone w Bibliotece dokumentów.


