Przejdź do
EP5895100007
Rotor for Eppendorf® 5920R Centrifuges
swing-bucket rotor, S-4x1000, aerosol-tight, to hold, 0.2 mL to 1,000 mL tubes and flasks (plate/tube buckets), with 4 × 1000 mL round buckets, with Eppendorf QuickLock® lid
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
Informacje o tej pozycji
NACRES:
NB.82
UNSPSC Code:
41103909
eCl@ss:
32110300
Manufacturer/tradename:
Eppendorf® 5895100007 (Global)
Compatibility:
for use with centrifuge models 5920R
Parameter:
3,428 × g max. RCF
Feature:
to hold 0.2 mL to 1,000 mL tubes and flasks (plate/tube buckets), aerosol-tight, swing-bucket rotor, with 4 × 1000 mL round buckets, with Eppendorf QuickLock® lid
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomóc
feature
to hold 0.2 mL to 1,000 mL tubes and flasks (plate/tube buckets), aerosol-tight, swing-bucket rotor, with 4 × 1000 mL round buckets, with Eppendorf QuickLock® lid
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5895100007 (Global)
parameter
3,428 × g max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5920R
General description
The large Centrifuge 5920 R is a benchtop centrifuge that combines exceptional capacity with a compact and ergonomic design. It has similar dimensions to refrigerated competitor models in the 3-liter segment, but has an outstanding capacity of up to 4 x 1,000 mL or 52 x 50 mL tubes. Optimize high-throughput centrifugation and make your daily work easier.
Legal Information
Eppendorf is a registered trademark of Eppendorf SE
Eppendorf QuickLock is a registered trademark of Eppendorf SE
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
EP5895101003EP5895190006EP5895135005
description
swing-bucket rotor, S-4x1000, aerosol-tight, to hold, 0.2 mL to 1,000 mL tubes and flasks (plate/tube buckets), with 4 × 1000 mL round buckets, with Eppendorf QuickLock® lid
description
swing-bucket rotor, S-4x1000, to hold, 4 x 1,000 mL buckets, without buckets
description
swing-bucket rotor, S-4x1000, aerosol-tight, with 4 x universal-large buckets
description
FA-48x2, aerosol tight, to hold, 48 x 1.5/2.0 mL tubes, with Eppendorf QuickLock rotor lid
compatibility
for use with centrifuge models 5920R
compatibility
for use with centrifuge models 5920R
compatibility
for use with centrifuge models 5910R
compatibility
for use with centrifuge models 5920R
manufacturer/tradename
Eppendorf® 5895100007 (Global)
manufacturer/tradename
Eppendorf® 5895101003 (Global)
manufacturer/tradename
Eppendorf® 5895190006 (Global)
manufacturer/tradename
Eppendorf® 5895135005 (Global)
feature
aerosol-tight, to hold 0.2 mL to 1,000 mL tubes and flasks (plate/tube buckets), swing-bucket rotor, with Eppendorf QuickLock® lid, with 4 × 1000 mL round buckets
feature
without buckets, swing-bucket rotor, to hold 4 x plate/tube buckets, to hold 4 x 1,000 mL buckets
feature
aerosol-tight, with 4 x universal-large buckets, swing-bucket rotor, to hold 0.2 to 750 mL ubes and flasks and plates
feature
aerosol tight, to hold 48 x 1.5/2.0 mL tubes, fixed angle rotor, with Eppendorf QuickLock rotor lid
technique(s)
centrifugation: suitable
technique(s)
centrifugation: suitable
technique(s)
centrifugation: suitable
technique(s)
centrifugation: suitable
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
parameter
3,428 × g max. RCF
parameter
3,153 × g max. RCF
parameter
4,200 × g max. RCF
parameter
20,913 × g max. RCF
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)