Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
material
red polypropylene (with biohazard warning symbol & precautions in English, Spanish, & French)
feature
label
packaging
pkg of 500 ea
manufacturer/tradename
Heathrow Scientific HS10320
H × W × thickness
305 mm × 203 mm × 0.04 mm
General description
A range of biohazard collection bags printed with biohazard warning symbol and precautions in English, Spanish, and French
- Includes an indicator patch that darkens when autoclaved
- Autoclavable to 250°F (121°C)
- Disposable
- Epoxy coated steel wire holder available HS10044H
Features and Benefits
- Two thicknesses of polypropylene are available
- An indicator patch that darkens when autoclaved is included
- Autoclavable to 250°F (121°C)
- Disposable
- An epoxy-coated steel wire holder is available (HS10044H).
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description H × W × thickness 305 mm × 203 mm × 0.04 mm, red polypropylene (with biohazard warning symbol & precautions in English, Spanish, & French) | description H × W × thickness 483 mm × 356 mm × 0.04 mm, red polypropylene (with biohazard warning symbol & precautions in English, Spanish, & French) | description H × W × thickness 584 mm × 483 mm × 0.04 mm, red polypropylene (with biohazard warning symbol & precautions in English, Spanish, & French) | description H × W × thickness 838 mm × 635 mm × 0.04 mm, red polypropylene (with biohazard warning symbol & precautions in English, Spanish, & French) |
| material red polypropylene (with biohazard warning symbol & precautions in English, Spanish, & French) | material red polypropylene (with biohazard warning symbol & precautions in English, Spanish, & French) | material red polypropylene (with biohazard warning symbol & precautions in English, Spanish, & French) | material red polypropylene (with biohazard warning symbol & precautions in English, Spanish, & French) |
| H × W × thickness 305 mm × 203 mm × 0.04 mm | H × W × thickness 483 mm × 356 mm × 0.04 mm | H × W × thickness 584 mm × 483 mm × 0.04 mm | H × W × thickness 838 mm × 635 mm × 0.04 mm |
| packaging pkg of 500 ea | packaging pkg of 200 ea | packaging pkg of 200 ea | packaging pkg of 200 ea |
| manufacturer/tradename Heathrow Scientific HS10320 | manufacturer/tradename Heathrow Scientific HS10321 | manufacturer/tradename Heathrow Scientific HS10322 | manufacturer/tradename Heathrow Scientific HS10323 |
| feature label | feature label | feature label | feature label |
Still not finding the right product?
Explore all of our products under Biohazard disposal bags
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?