Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
material
polypropylene
feature
Holds 432 x 5-7 mL tubes
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS120112
size
673x235x90 mm
color
white
General description
- Fully-assembled, polypropylene Mega Tube Racks are great for high throughput labs requiring large sample storage.
- Sturdy and light weight material for easy transport
- Alphanumeric identification for quick location and retrieval of tubes
- Place different test groupings into one rack and eliminate need for multiple tube racks
- Autoclavable
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description double, for 10-13 mm tubes, white | description double, for 10-13 mm tubes, blue | description double, for 13-16 mm tubes, white | description single, for 10-13 mm tubes, white |
| feature Holds 432 x 5-7 mL tubes | feature Holds 432 x 5-7 mL tubes | feature Holds 240 x 10 mL tubes | feature Holds 216 x 5-7 mL tubes |
| manufacturer/tradename Heathrow Scientific HS120112 | manufacturer/tradename Heathrow Scientific HS120113 | manufacturer/tradename Heathrow Scientific HS120114 | manufacturer/tradename Heathrow Scientific HS24322E |
| material polypropylene | material polypropylene | material polypropylene | material polypropylene |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging - |
| color white | color blue | color white | color white |
| size 673x235x90 mm | size 673x235x90 mm | size 673x267x90 mm | size 337x235x91 mm |
Still not finding the right product?
Explore all of our products under Mega Rack® tube rack
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?