Przejdź do
HS24322E
Mega Rack® tube rack
single, for 10-13 mm tubes, white
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
Informacje o tej pozycji
NACRES:
NB.12
UNSPSC Code:
41122804
eCl@ss:
32032201
Manufacturer/tradename:
Heathrow Scientific HS24322E
Feature:
Holds 216 x 5-7 mL tubes
Material:
polypropylene
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomóc
material
polypropylene
feature
Holds 216 x 5-7 mL tubes
manufacturer/tradename
Heathrow Scientific HS24322E
size
337x235x91 mm
color
white
General description
Mega Rack High-capacity storage rack for sample and blood collection tubes.
  • Fully-assembled, polypropylene Mega Tube Racks are great for high throughput labs requiring large sample storage.
  • Sturdy and light weight material for easy transport
  • Alphanumeric identification for quick location and retrieval of tubes
  • Place different test groupings into one rack and eliminate need for multiple tube racks
  • Autoclavable
Legal Information
Mega Rack is a registered trademark of Heathrow Scientific
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
HS24323EHS120112HS24322A
description
single, for 10-13 mm tubes, white
description
single, for 13-16 mm tubes, white
description
double, for 10-13 mm tubes, white
description
single, for 10-13 mm tubes, blue
feature
Holds 216 x 5-7 mL tubes
feature
Holds 120 x 10 mL tubes
feature
Holds 432 x 5-7 mL tubes
feature
Holds 216 x 5-7 mL tubes
manufacturer/tradename
Heathrow Scientific HS24322E
manufacturer/tradename
Heathrow Scientific HS24323E
manufacturer/tradename
Heathrow Scientific HS120112
manufacturer/tradename
Heathrow Scientific HS24322A
material
polypropylene
material
polypropylene
material
polypropylene
material
polypropylene
size
337x235x91 mm
size
337x235x91 mm
size
673x235x90 mm
size
337x235x91 mm
color
white
color
white
color
white
color
blue
Still not finding the right product?
Explore all of our products under Mega Rack® tube rack
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)