Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
72 trays | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 3410,00 zł |
Informacje o tej pozycji
3410,00 zł
material
cardboard , yellow (trim)
feature
Holds 20 x slides 76x26 mm
packaging
case of 72 trays
manufacturer/tradename
Heathrow Scientific HS120197
L × W × H
208 × 341 × 9 mm
Features and Benefits
- Easily organize slides with the index on the front cover that corresponds with the number grid inside
- Recessed slide compartments and inside covers protect specimens. Slides cannot be touched when the cover is closed, reducing contamination
- Front protection flaps are attached with durable tape for repeated use. Fold flaps flat under the slide holder to save space when in use
- Thumb cut in slide compartments enables easy removal of slides with slight pressure at one end
- Compartment holder is made from a double layer of densely pressed cardboard. Cover is a single layer cardboard
- Disposable
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description Holds 20 x slides 76x26 mm, yellow (trim) | description Holds 20 x slides 76x26 mm, black (trim) | description 100 place, red | description 100 place, blue |
| manufacturer/tradename Heathrow Scientific HS120197 | manufacturer/tradename Heathrow Scientific HS9902 | manufacturer/tradename Heathrow Scientific HS15991B | manufacturer/tradename Heathrow Scientific HS15991A |
| feature Holds 20 x slides 76x26 mm | feature Holds 20 x slides 76x26 mm | feature Holds 100 x slides 76x26 mm | feature Holds 100 x slides 76x26 mm |
| material yellow (trim), cardboard | material black (trim) | material polypropylene | material polypropylene |
| packaging case of 72 trays | packaging case of 72 trays | packaging - | packaging - |
| L × W × H 208 × 341 × 9 mm | L × W × H 208 × 341 × 9 mm | L × W × H - | L × W × H - |
Still not finding the right product?
Explore all of our products under Cardboard Slide Tray
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?