Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
material
white polystyrene
sterility
sterile; electron beam irradiated
feature
to hold round pills and capsules
packaging
pack of 30 ea (3 x packs of 10 individually wrapped)
manufacturer/tradename
Heathrow Scientific 120847
L × W × H
23.0 in. × 23.0 in. × 1.3 in.
General description
Intended for single use to prevent cross-contamination for batch sampling or distribution of pills and capsules.
For Round Pills.
Available in both, sterile and non-sterile for intended usage.
Features and Benefits
- Disposable to ensure no chemical cross-contamination for patient Safety
- Made from FDA food grade polystyrene in an ISO Class 8 clean room
- Meets quality control and GMP requirements for manufacturing batch sampling
- Durable and shatterproof trays constructed of inert, BSE/TSE free material
Convenience
- Pour spout in corner for dispensing control
- Ergonomic spatula (length - 6.9 in/17.6 cm)
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description sterile; electron beam irradiated | description non-sterile | description sterile; electron beam irradiated | description non-sterile |
| material white polystyrene | material white polystyrene | material white polystyrene | material white polystyrene |
| packaging pack of 30 ea (3 x packs of 10 individually wrapped) | packaging pack of 30 ea (3 x packs of 10 ea) | packaging pack of 10 ea | packaging pack of 10 ea |
| feature to hold round pills and capsules | feature to hold round pills and capsules | feature to hold size 00 to 2 tablets and capsules | feature to hold size 00 to 2 tablets and capsules |
| L × W × H 23.0 in. × 23.0 in. × 1.3 in. | L × W × H 23.0 in. × 23.0 in. × 1.3 in. | L × W × H 10.3 in. × 5.2 in. × 0.5 in. | L × W × H 10.3 in. × 5.2 in. × 0.5 in. |
| manufacturer/tradename Heathrow Scientific 120847 | manufacturer/tradename Heathrow Scientific 120848 | manufacturer/tradename Heathrow Scientific 120845 | manufacturer/tradename Heathrow Scientific 120846 |
| sterility sterile; electron beam irradiated | sterility non-sterile | sterility sterile; electron beam irradiated | sterility non-sterile |
Still not finding the right product?
Explore all of our products under Triangular Pill Counting Tray
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?