Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
material
acrylic , green (dispenser)
feature
2 x 50 mm or 1 x 100 mm Parafilm M Sealing Film roll
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS234525C
L × W × H
120 mm × 156 mm × 171 mm
General description
Features and Benefits
- ABS plastic dispenser designed with a safety razor that slides and cuts with no danger to fingers.
- Supplied with two easily exchanged cutting blades
- Dispenser holds one or two 50 mm (2-inch) rolls, or one 100 mm (4-inch) roll
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description green (dispenser) | description blue (dispenser) | description clear (dispenser) | description blue (dispenser) |
| manufacturer/tradename Heathrow Scientific HS234525C | manufacturer/tradename Heathrow Scientific HS234525B | manufacturer/tradename Heathrow Scientific HS234524 | manufacturer/tradename Heathrow Scientific 120667 |
| material green (dispenser), acrylic | material blue (dispenser), acrylic | material acrylic , clear (dispenser) | material blue (dispenser), acrylonitrile-butadiene-styrene (ABS) |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea |
| feature 2 x 50 mm or 1 x 100 mm Parafilm M Sealing Film roll | feature holds 2 x 50 mm or 1 x 100 mm Parafilm M Sealing Film roll | feature holds 2 x 50 mm or 1 x 100 mm ParafilmM Sealing Film roll | feature to hold 2 x 50 mm or 1 x 100 mm Parafilm M Sealing Film roll |
| L × W × H 120 mm × 156 mm × 171 mm | L × W × H 120 mm × 156 mm × 171 mm | L × W × H 119 mm × 168 mm × 175 mm | L × W × H 123 mm × 172 mm × 99 mm |
Still not finding the right product?
Explore all of our products under Parafilm® Sealing Film Dispenser, Blue
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?