Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
2 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 164,00 zł |
Informacje o tej pozycji
164,00 zł
material
blue , fiberglass , polypropylene (reinforced)
feature
to hold 50 tube(s) 14-17 mm tubes
packaging
pack of 2 ea
manufacturer/tradename
Heathrow Scientific HS24311B
General description
Features and Benefits
- Constructed of fiberglass-reinforced polypropylene
- Spacious peg design allows easy viewing of tube contents
- Use as a drying rack for glass plates or upside-down tubes
- Pegs remain straight even after repeated autoclaving
- Embossed alphanumeric identification for quick location and retrieval of tubes
- Unique tear drop well design allows for better retention of tubes
- Autoclavable
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description to hold, 50 tube(s) 14-17 mm tubes, blue | description to hold, 50 tube(s) 14-17 mm tubes, white | description to hold, 96 tube(s) 10-13 mm tubes, white | description to hold, 96 tube(s) 10-13 mm tubes, blue |
| feature to hold 50 tube(s) 14-17 mm tubes | feature to hold 50 tube(s) 14-17 mm tubes | feature to hold 96 tube(s) 10-13 mm tubes | feature to hold 96 tube(s) 10-13 mm tubes |
| manufacturer/tradename Heathrow Scientific HS24311B | manufacturer/tradename Heathrow Scientific HS24311A | manufacturer/tradename Heathrow Scientific HS24312A | manufacturer/tradename Heathrow Scientific HS24312B |
| material blue , fiberglass , polypropylene (reinforced) | material polypropylene (reinforced), white , fiberglass | material polypropylene , fiberglass , white | material polypropylene , fiberglass , blue |
| packaging pack of 2 ea | packaging pack of 2 ea | packaging pack of 2 ea | packaging pack of 2 ea |
Still not finding the right product?
Explore all of our products under Peg Tube Rack
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?