Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
material
white , fiberglass , polypropylene
feature
to hold 96 tube(s) 10-13 mm tubes
packaging
pack of 2 ea
manufacturer/tradename
Heathrow Scientific HS24312A
L × W × H
82 mm × 126 mm × 69 mm
General description
Features and Benefits
Spacious peg design allows easy viewing of tube contents
Use as a drying rack for glass plates or upside-down tubes
Pegs remain straight even after repeated autoclaving
Embossed alphanumeric identification for quick location and retrieval of tubes
Unique tear drop well design allows for better retention of tubes
Autoclavable
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description to hold, 96 tube(s) 10-13 mm tubes, white | description to hold, 96 tube(s) 10-13 mm tubes, blue | description to hold, 50 tube(s) 14-17 mm tubes, white | description to hold, 50 tube(s) 14-17 mm tubes, blue |
| feature to hold 96 tube(s) 10-13 mm tubes | feature to hold 96 tube(s) 10-13 mm tubes | feature to hold 50 tube(s) 14-17 mm tubes | feature to hold 50 tube(s) 14-17 mm tubes |
| manufacturer/tradename Heathrow Scientific HS24312A | manufacturer/tradename Heathrow Scientific HS24312B | manufacturer/tradename Heathrow Scientific HS24311A | manufacturer/tradename Heathrow Scientific HS24311B |
| material white , polypropylene , fiberglass | material fiberglass , polypropylene , blue | material fiberglass , white , polypropylene (reinforced) | material fiberglass , polypropylene (reinforced), blue |
| packaging pack of 2 ea | packaging pack of 2 ea | packaging pack of 2 ea | packaging pack of 2 ea |
| L × W × H 82 mm × 126 mm × 69 mm | L × W × H 82 mm × 126 mm × 69 mm | L × W × H 188 mm × 104 mm × 69 mm | L × W × H - |
Still not finding the right product?
Explore all of our products under Peg Tube Rack
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?