Przejdź do zawartości
Merck

Przejdź do

MIR270

TransIT®-293 Transfection Reagent

A high efficiency, low toxicity, DNA transfection reagent optimized for 293 cells

Synonim(y):

HEK 293 cell transfection, MIR 270

Zaloguj się, aby wyświetlić ceny organizacyjne i kontraktowe.

Wybierz wielkość

Zmień widok
Rozmiar/SKUDostępnośćCena netto

Informacje o tej pozycji

UNSPSC Code:
41106502
Form:
liquid
Grade:
Molecular Biology
Technique(s):
transfection: suitable
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Potrzebujesz pomocy? Nasz zespół doświadczonych naukowców chętnie Ci pomoże.
Pozwól nam pomóc


Nazwa produktu

TransIT®-293, A high efficiency, low toxicity, DNA transfection reagent optimized for 293 cells

grade

Molecular Biology

form

liquid

manufacturer/tradename

Mirus Bio

storage condition

do not freeze

technique(s)

transfection: suitable

shipped in

wet ice

storage temp.

2-8°C

General description

TransIT®-293 Transfection Reagent is specifically optimized to provide exceptional transfection efficiency of plasmid DNA in HEK 293 and cell types of associated lineage. TransIT®-293 provides all the attributes of the trusted TransIT series of transfection reagents: high transfection efficiency, low toxicity, serum compatibility, simplicity of use and reproducibility.
Transfections with TransIT®-293 Reagent do not require medium changes and can be carried out in serum-containing medium. TransIT®-293 is suitable for both transient and stable transfection and can be used for many applications such as gene expression, viral production, shRNA expression and promoter analysis.

Features and Benefits

  • Unique Formulation – Maximize transfection performance in HEK 293 cells
  • High Efficiency Delivery – Achieve 75-85% transfection efficiency to ensure experimental success
  • Deliver Single or Multiple Plasmids – Ideal for specialized applications, such as virus production
  • Low Cellular Toxicity – Maintain cell density and reduce experimental biases

Other Notes

Biological Source: raw materials that are derived from animals.

Suitable for: HEK 293 transfection

Legal Information

TransIT is a registered trademark of Mirus Bio
Ta strona może zawierać tekst przetłumaczony maszynowo.

Porównaj podobne pozycje

Wyświetl pełne porównanie

Pokaż różnice

1 of 1

Ta pozycja
MIR230MIR57MIR540
TransIT®-293 A high efficiency, low toxicity, DNA transfection reagent optimized for 293 cells

MIR270

TransIT®-293

TransIT®-LT1 A low toxicity, broad spectrum DNA transfection reagent

MIR230

TransIT®-LT1

TransIT®-PRO High, Reproducible Protein Yield in Suspension CHO and 293 Cells

MIR57

TransIT®-PRO

TransIT®-2020 A high performance, animal-free, broad spectrum DNA transfection reagent

MIR540

TransIT®-2020

description

A high efficiency, low toxicity, DNA transfection reagent optimized for 293 cells

description

A low toxicity, broad spectrum DNA transfection reagent

description

High, Reproducible Protein Yield in Suspension CHO and 293 Cells

description

A high performance, animal-free, broad spectrum DNA transfection reagent

technique(s)

transfection: suitable

technique(s)

transfection: suitable

technique(s)

transfection: suitable

technique(s)

transfection: suitable

grade

Molecular Biology

grade

Molecular Biology

grade

Molecular Biology

grade

Molecular Biology

form

liquid

form

liquid

form

liquid

form

liquid

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

storage condition

do not freeze

storage condition

-

storage condition

-

storage condition

-

manufacturer/tradename

Mirus Bio

manufacturer/tradename

Mirus Bio

manufacturer/tradename

Mirus Bio

manufacturer/tradename

Mirus Bio




Wybierz jedną z najnowszych wersji:

Certyfikaty analizy (CoA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumenty section.

Proszę o kontakt, jeśli potrzebna jest pomoc Obsługa Klienta

Masz już ten produkt?

Dokumenty związane z niedawno zakupionymi produktami zostały zamieszczone w Bibliotece dokumentów.

Odwiedź Bibliotekę dokumentów