Przejdź do
Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
Nazwa produktu
TransIT®-PRO, High, Reproducible Protein Yield in Suspension CHO and 293 Cells
grade
Molecular Biology
form
liquid
manufacturer/tradename
Mirus Bio
technique(s)
transfection: suitable
shipped in
wet ice
storage temp.
−20°C
General description
Features and Benefits
- High Protein Yield – in Suspension CHO and 293 cells, including CHO-S and Expi293F cells
- Compatible with Multiple CHO Media Formulations – Use with CHOgro Expression Medium to Achieve Highest Yields
- Simple Workflow – Reproducible Protein Expression with Minimal Optimization
Packaging
pkg of 10 mL, without PRO Boost (MIR5750)
Other Notes
Suitable for:
- CHO transfection
- HEK 293 transfection
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description High, Reproducible Protein Yield in Suspension CHO and 293 Cells | description A high efficiency, low toxicity, DNA transfection reagent optimized for 293 cells | description A low toxicity, broad spectrum DNA transfection reagent | description A high efficiency, low toxicity transfection reagent for large RNA and CRISPR guide RNA |
| technique(s) transfection: suitable | technique(s) transfection: suitable | technique(s) transfection: suitable | technique(s) transfection: suitable |
| grade Molecular Biology | grade Molecular Biology | grade Molecular Biology | grade Molecular Biology |
| form liquid | form liquid | form liquid | form liquid |
| storage temp. −20°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
| shipped in wet ice | shipped in wet ice | shipped in wet ice | shipped in wet ice |
| manufacturer/tradename Mirus Bio | manufacturer/tradename Mirus Bio | manufacturer/tradename Mirus Bio | manufacturer/tradename Mirus Bio |
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?
Dokumenty związane z niedawno zakupionymi produktami zostały zamieszczone w Bibliotece dokumentów.



