Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
material
clear (blue printed warnings), polypropylene
packaging
pack of 200 ea (bulk bag)
manufacturer/tradename
MTC™ Bio A9000C
W × H
12 in. (31 cm) × 26 in. (66 cm)
General description
Features and Benefits
- Heavy gauge polypropylene
- Large white marking areas
- Strong seamless bottom
- Printed biohazard symbol
- Withstands autoclaving at temperatures up to 134˚C (270˚F)
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description W × H 12 in. (31 cm) × 26 in. (66 cm), clear (blue printed warnings) | description W × H 12 in. (31 cm) × 26 in. (66 cm), red (black printed warnings), pack of 200 ea (bulk bag) | description W × H 24 in. (61 cm) × 32 in. (81 cm), red (black printed warnings) | description W × H 24 in. (61 cm) × 32 in. (81 cm), clear (blue printed warnings) |
| material polypropylene , clear (blue printed warnings) | material polypropylene , red (black printed warnings) | material polypropylene , red (black printed warnings) | material polypropylene , clear (blue printed warnings) |
| W × H 12 in. (31 cm) × 26 in. (66 cm) | W × H 12 in. (31 cm) × 26 in. (66 cm) | W × H 24 in. (61 cm) × 32 in. (81 cm) | W × H 24 in. (61 cm) × 32 in. (81 cm) |
| manufacturer/tradename MTC™ Bio A9000C | manufacturer/tradename MTC™ Bio A9000R | manufacturer/tradename MTC™ Bio A9002R | manufacturer/tradename MTC™ Bio A9002C |
| packaging pack of 200 ea (bulk bag) | packaging pack of 200 ea (bulk bag) | packaging pack of 200 ea (bulk bag) | packaging pack of 200 ea (bulk bag) |
Still not finding the right product?
Explore all of our products under MTC™ Bio Autoclave Bags
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?