Przejdź do
MTCA9002R
MTC™ Bio Autoclave Bags
W × H 24 in. (61 cm) × 32 in. (81 cm), red (black printed warnings)
Synonim(y):
biohazard bag, Autoclave pouches, MTC Bio A9002R, Steam sterilization bags, sterilization bag
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
Informacje o tej pozycji
NACRES:
NB.15
UNSPSC Code:
24111500
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomóc
material
polypropylene , red (black printed warnings)
packaging
pack of 200 ea (bulk bag)
manufacturer/tradename
MTC™ Bio A9002R
W × H
24 in. (61 cm) × 32 in. (81 cm)
General description
These high-temperature autoclavable bags are manufactured from heavy-gauge polypropylene material that withstands autoclaving at temperatures up to 134˚C (270˚F). Their ability to contain is completely unaffected at standard autoclave temperature (121˚C). Each bag has an oversized white marking panel, suitable for use with all types of permanent markers. They are also prominently marked with the words “Hazardous Waste” and the standard “Biohazard” symbol, conforming to the OSHA standard (29CFR1910.1030). Designed to resist stretching and poking, these bags are extremely pliable, leak-resistant and have seamless bottoms for exceptional strength and integrity. Both sizes (12×26 inch and 24×32 inch) are available in clear or opaque “biohazard” red color. They are supplied in units of 200 bags, with 4 inner packs of 50 bags for convenient dispensing and storage.
Autoclave bags, 24x32" (61 x 81.3cm), red, biohazard, printed, marking area, 200/cs
Features and Benefits
  • Heavy gauge polypropylene
  • Large white marking areas
  • Strong seamless bottom
  • Printed biohazard symbol
  • Withstands autoclaving at temperatures up to 134˚C (270˚F)
Legal Information
MTC is a trademark of MTC Bio
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
MTCA9002CMTCA9004RMTCA9000R
description
W × H 24 in. (61 cm) × 32 in. (81 cm), red (black printed warnings)
description
W × H 24 in. (61 cm) × 32 in. (81 cm), clear (blue printed warnings)
description
W × H 8.5 in. (21.6 cm) × 11 in. (27.9 cm), red (black printed warnings)
description
W × H 12 in. (31 cm) × 26 in. (66 cm), red (black printed warnings), pack of 200 ea (bulk bag)
material
red (black printed warnings), polypropylene
material
polypropylene , clear (blue printed warnings)
material
polypropylene , red (black printed warnings)
material
red (black printed warnings), polypropylene
W × H
24 in. (61 cm) × 32 in. (81 cm)
W × H
24 in. (61 cm) × 32 in. (81 cm)
W × H
8.5 in. (21.6 cm) × 11 in. (27.9 cm)
W × H
12 in. (31 cm) × 26 in. (66 cm)
manufacturer/tradename
MTC™ Bio A9002R
manufacturer/tradename
MTC™ Bio A9002C
manufacturer/tradename
MTC™ Bio A9004R
manufacturer/tradename
MTC™ Bio A9000R
packaging
pack of 200 ea (bulk bag)
packaging
pack of 200 ea (bulk bag)
packaging
pack of 100 ea (bulk bag)
packaging
pack of 200 ea (bulk bag)
Still not finding the right product?
Explore all of our products under MTC™ Bio Autoclave Bags
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)