Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
material
polystyrene
packaging
pack of 4 ea
manufacturer/tradename
MTC™ Bio B1206
L × W × D
33 mm (4 in.) × 103 mm (1 1/4 in.) × 35 mm (1 3/8 in.)
General description
Features and Benefits
- A perfect box for every blot
- Reduce antibody usage
- Hinged lid minimizes evaporation
- For halves, wholes, strips or minis
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description 6 compartments, L × W × D 33 mm (4 in.) × 103 mm (1 1/4 in.) × 35 mm (1 3/8 in.) | description 4 compartments, L × W × D 32 mm (11/4 in.) × 52 mm (2 5/16 in.) × 28 mm (1 1/8 in.), for various gels | description 4 compartments, L × W × D 38 mm (1 1/2 in.) × 71 mm (2 13/16 in.) × 28 mm (1 1/8 in.), for various gels including half-mini protein gels | description 2 compartments, L × W × D 85 mm (311/32 in.) × 85 mm (311/32 in.) × 30 mm (13/16 in.), for Novex Mini (8x8cm), Bio-Rad Mini (7x8cm) and others |
| manufacturer/tradename MTC™ Bio B1206 | manufacturer/tradename MTC™ Bio B1203 | manufacturer/tradename MTC™ Bio B1204 | manufacturer/tradename MTC™ Bio B1212 |
| material polystyrene | material polystyrene | material polystyrene | material polystyrene |
| packaging pack of 4 ea | packaging pack of 6 ea | packaging pack of 6 ea | packaging pack of 6 ea |
| L × W × D 33 mm (4 in.) × 103 mm (1 1/4 in.) × 35 mm (1 3/8 in.) | L × W × D 32 mm (11/4 in.) × 52 mm (2 5/16 in.) × 28 mm (1 1/8 in.) | L × W × D 38 mm (1 1/2 in.) × 71 mm (2 13/16 in.) × 28 mm (1 1/8 in.) | L × W × D 85 mm (311/32 in.) × 85 mm (311/32 in.) × 30 mm (13/16 in.) |
Still not finding the right product?
Explore all of our products under MTC™ Bio MultiBox™ for Western Blotting
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?