Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
1000 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 1410,00 zł |
Informacje o tej pozycji
1410,00 zł
material
(self standing star foot), cap (screw top ), clear , polypropylene
sterility
sterile
feature
cap natural, neck thread external
packaging
pack of 1000 ea (20 x bags 50 ea)
manufacturer/tradename
MTC™ Bio C3220-SG
parameter
22,000 × g speed
capacity
2.0 mL
diam. × L
13.1 mm × 46.8 mm
General description
Features and Benefits
- Ultra-clear polypropylene
- Packaged in resealable bags of 50
- Leak-proof O-ring seal
- Sterile, RNase, DNase & Pyrogen free
- White, printed graduations
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description capacity 2.0 mL, sterile, cap, natural, self standing, pack of 1000 ea (20 x bags 50 ea) | description capacity 2.0 mL, sterile, cap, natural(looped), self standing, pack of 1000 ea (20 x bags 50 ea) | description capacity 2.0 mL, sterile, cap, natural, conical bottom, pack of 1000 ea (20 x bags 50 ea) | description capacity 1.5 mL, sterile, cap, natural, self standing, pack of 1000 ea (20 x bags 50 ea) |
| manufacturer/tradename MTC™ Bio C3220-SG | manufacturer/tradename MTC™ Bio C3220-SGL | manufacturer/tradename MTC™ Bio C3220-CG | manufacturer/tradename MTC™ Bio C3215-SG |
| parameter 22,000 × g speed | parameter 22,000 × g speed | parameter 22,000 × g speed | parameter 22,000 × g speed |
| sterility sterile | sterility sterile | sterility sterile | sterility sterile |
| material cap (screw top ), clear , (self standing star foot), polypropylene | material (self standing star foot), clear , cap (screw top loop ), polypropylene | material screw top cap, clear , conical bottom, round , polypropylene | material screw top cap, clear , round , polypropylene |
| capacity 2.0 mL | capacity 2.0 mL | capacity 2.0 mL | capacity 1.5 mL |
| packaging pack of 1000 ea (20 x bags 50 ea) | packaging pack of 1000 ea (20 x bags 50 ea) | packaging pack of 1000 ea (20 x bags 50 ea) | packaging pack of 1000 ea (20 x bags 50 ea) |
Still not finding the right product?
Explore all of our products under MTC™ Bio ClearSeal™ Graduated Screw Cap Microtubes
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?