Przejdź do zawartości
Merck

Przejdź do

MTCD309627S

MTC™ Bio OptiWell™ Deep Well Plate

96 round wells, U-bottom wells, capacity 1.2 mL, sterile, case of 50 ea (10 bags of 5ea)

Synonim(y):

automated research plate, MTC Bio D3096-27S, automated assay plate, lab automation plate, research automation tray, robotic research plate

Zaloguj się, aby wyświetlić ceny organizacyjne i kontraktowe.

Wybierz wielkość

Zmień widok
Rozmiar/SKUDostępnośćCena netto

Informacje o tej pozycji

UNSPSC Code:
41121800
NACRES:
NB.15
eCl@ss:
32039090
Material:
U-bottom wells, clear plate, clear wells, polypropylene , round wells
Size:
96 wells
Sterility:
sterile
Feature:
lid: no, skirt: no
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Potrzebujesz pomocy? Nasz zespół doświadczonych naukowców chętnie Ci pomoże.
Pozwól nam pomóc


material

U-bottom wells, clear plate, clear wells, polypropylene , round wells

sterility

sterile

feature

lid: no, skirt: no

packaging

case of 50 ea (10 bags of 5ea)

manufacturer/tradename

MTC™ Bio D3096-27S

parameter

4,000 × g speed

capacity

1.2 mL

size

96 wells

well volume

1.2 mL

General description

OptiWell plates come in 24, 48, 96 or 384 format for virtually every research lab, HTS application and automated/robotic platform. These plates are useful for bacterial cultures, centrifugation (up to 4000 x g) and general sample storage. Plates are produced/packaged in a Class 100,000 clean room and are ANSI / SLAS 1-2004 compliant. Several plates are sterilized by gamma irradiation and all are certified non-pyrogenic, RNase/DNase free.

Features and Benefits

  • For automated research platforms / robotic equipment
  • Produced/packaged in a Class 100,000 clean room
  • ANSI / SLAS 1-2004 compliant
  • Minimal binding / liquid retention
  • Black printed alpha-numerics
  • Virgin, medical-grade polypropylene
  • Sterilized by gamma irradiation
  • Certified non-pyrogenic, RNase/DNase free

Legal Information

MTC is a trademark of MTC Bio
OptiWell is a trademark of MTC Bio
Ta strona może zawierać tekst przetłumaczony maszynowo.

Porównaj podobne pozycje

Wyświetl pełne porównanie

Pokaż różnice

1 of 1

Ta pozycja
MTCD309627MTCD309625MTCD309615
description

96 round wells, U-bottom wells, capacity 1.2 mL, sterile, case of 50 ea (10 bags of 5ea)

description

96 round wells, U-bottom wells, capacity 1.2 mL, non-sterile, case of 50 ea (10 bags of 5ea)

description

96 square wells, U-bottom wells, capacity 1.2 mL, non-sterile, case of 50 ea (10 bags of 5ea)

description

96 round wells, U-bottom wells, capacity 1.0 mL, non-sterile, case of 50 ea (10 bags of 5ea)

manufacturer/tradename

MTC™ Bio D3096-27S

manufacturer/tradename

MTC™ Bio D3096-27

manufacturer/tradename

MTC™ Bio D3096-25

manufacturer/tradename

MTC™ Bio D3096-15

feature

lid: no, skirt: no

feature

skirt: no, lid: no

feature

lid: no, skirt: no

feature

lid: no, skirt: no

sterility

sterile

sterility

non-sterile

sterility

non-sterile

sterility

non-sterile

material

U-bottom wells, clear plate, polypropylene , clear wells, round wells

material

polypropylene , round wells, clear plate, clear wells, U-bottom wells

material

clear wells, clear plate, U-bottom wells, square wells, polypropylene

material

U-bottom wells, clear wells, round wells, clear plate, polypropylene

size

96 wells

size

96 wells

size

96 wells

size

96 wells

packaging

case of 50 ea (10 bags of 5ea)

packaging

case of 50 ea (10 bags of 5ea)

packaging

case of 50 ea (10 bags of 5ea)

packaging

case of 50 ea (10 bags of 5ea)


Still not finding the right product?




Wybierz jedną z najnowszych wersji:

Certyfikaty analizy (CoA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumenty section.

Proszę o kontakt, jeśli potrzebna jest pomoc Obsługa Klienta

Masz już ten produkt?

Dokumenty związane z niedawno zakupionymi produktami zostały zamieszczone w Bibliotece dokumentów.

Odwiedź Bibliotekę dokumentów