Przejdź do
MTCF4061F
MTC™ Bio BioFlask™ Erlenmeyer Shaker Flask, Individually Wrapped
capacity 250 mL, polypropylene, flat bottom, cap, vented, sterile
Synonim(y):
culture flask, MTC Bio F4061-F, laboratory flask, plastic flask, shaker flask
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
Informacje o tej pozycji
NACRES:
NB.15
UNSPSC Code:
41122104
Capacity:
250 mL
Material:
flat bottom, polypropylene
Sterility:
sterile
Feature:
cap vented
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomóc
material
flat bottom, polypropylene
sterility
sterile
feature
cap vented
packaging
pack of 12 ea (individually wrapped)
manufacturer/tradename
MTC™ Bio F4061-F
capacity
250 mL
General description
Erlenmeyer Shaker Flasks are made of polycarbonate or PETG. The polypropylene screw caps are vented with 0.2µm membranes to increase air transmission and enhance cell growth. The mouth size conforms with the international SBS standard, for quick, secure connection to bioreactor equipment. Polycarbonate flasks have baffled (ribbed) bottoms to provide increased oxygenation and mixing efficiency even at lower shaking speeds. Polycarbonate Flasks are autoclavable at 121°C and 15 psi. All flasks, regardless of size, are individually wrapped and gamma sterilized to SAL 10-6. They are free of RNase, DNase, pyrogen, endotoxins and human DNA.
Flask, Erlenmeyer, 250ml, PETG, flat bottom w/ vented screw cap, Sterile, indiv wrapped, 12/CS
Features and Benefits
  • Vented screw caps
  • Flat or baffled bottoms
  • Gamma sterile (SAL 10-6)
  • Pyrogen free and RNase/DNase free
  • Individually wrapped
Legal Information
BioFlask is a trademark of MTC Bio
MTC is a trademark of MTC Bio
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
MTCF4061BMTCF4060FMTCF4062B
description
capacity 250 mL, polypropylene, flat bottom, cap, vented, sterile
description
capacity 250 mL, polycarbonate, bottom (baffled), cap, vented, sterile
description
capacity 125 mL, polypropylene, flat bottom, cap, vented, sterile
description
capacity 500 mL, polycarbonate, bottom (baffled), cap, vented, sterile
manufacturer/tradename
MTC™ Bio F4061-F
manufacturer/tradename
MTC™ Bio F4061-B
manufacturer/tradename
MTC™ Bio F4060-F
manufacturer/tradename
MTC™ Bio F4062-B
sterility
sterile
sterility
sterile
sterility
sterile
sterility
sterile
capacity
250 mL
capacity
250 mL
capacity
125 mL
capacity
500 mL
material
polypropylene , flat bottom
material
bottom (baffled), polycarbonate
material
flat bottom, polypropylene
material
polycarbonate , bottom (baffled)
feature
cap vented
feature
cap vented
feature
cap vented
feature
cap vented
packaging
pack of 12 ea (individually wrapped)
packaging
pack of 12 ea (individually wrapped)
packaging
pack of 24 ea (individually wrapped)
packaging
pack of 12 ea (individually wrapped)
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)